missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LRRC14 Full-length ORF (NP_055480.1, 1 a.a. - 493 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009684-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136198

  • 6845.00 SEK / 25µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MHTLVFLSTRQVLQCQPAACQALPLLPRELFPLLFKVAFMDKKTVVLRELVHTWPFPLLSFQQLLQECAHCSRALLQERPSTESMQAVILGLTARLHTSEPGASTQPLCRKHALRVLDMTGLLDDGVEQDPGTMSMWDCTAAVARTCIAQQQGGAAEPGPAPIPVEVRVDLRVNRASYAFLREALRSSVGSPLRLCCRDLRAEDLPMRNTVALLQLLDAGCLRRVDLRFNNLGLRGLSVIIPHVARFQHLASLRLHYVHGDSRQPSVDGEDNFRYFLAQMGRFTCLRELSMGSSLLSGRLDQLLSTLQSPLESLELAFCALLPEDLRFLARSPHAAHLKKLDLSGNDLSGSQLAPFQGLLQASAATLLHLELTECQLADTQLLATLPILTQCASLRYLGLYGNPLSMAGLKELLRDSVAQAELRTVVHPFPVDCYEGLPWPPPASVLLEASINEEKFARVEAELHQLLLASGRAHVLWTTDIYGRLAADYFSL
Specifications

Specifications

NP_055480.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
80.9kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0014
LRRC14
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9684
LRRC14 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MHTLVFLSTRQVLQCQPAACQALPLLPRELFPLLFKVAFMDKKTVVLRELVHTWPFPLLSFQQLLQECAHCSRALLQERPSTESMQAVILGLTARLHTSEPGASTQPLCRKHALRVLDMTGLLDDGVEQDPGTMSMWDCTAAVARTCIAQQQGGAAEPGPAPIPVEVRVDLRVNRASYAFLREALRSSVGSPLRLCCRDLRAEDLPMRNTVALLQLLDAGCLRRVDLRFNNLGLRGLSVIIPHVARFQHLASLRLHYVHGDSRQPSVDGEDNFRYFLAQMGRFTCLRELSMGSSLLSGRLDQLLSTLQSPLESLELAFCALLPEDLRFLARSPHAAHLKKLDLSGNDLSGSQLAPFQGLLQASAATLLHLELTECQLADTQLLATLPILTQCASLRYLGLYGNPLSMAGLKELLRDSVAQAELRTVVHPFPVDCYEGLPWPPPASVLLEASINEEKFARVEAELHQLLLASGRAHVLWTTDIYGRLAADYFSL
RUO
LRRC14
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers