Learn More
Abnova™ Human LUZP5 Partial ORF (NP_060230, 177 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00054892-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Condensin complexes I and II play essential roles in mitotic chromosome assembly and segregation. Both condensins contain 2 invariant structural maintenance of chromosome (SMC) subunits, SMC2 (MIM 605576) and SMC4 (MIM 605575), but they contain different sets of non-SMC subunits. NCAPG2 is 1 of 3 non-SMC subunits that define condensin II (Ono et al., 2003 [PubMed 14532007]).[supplied by OMIM]
Sequence: TKTGADVCRLWRIHQALYCFDYDLEESGEIKDMLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKASSpecifications
NP_060230 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TKTGADVCRLWRIHQALYCFDYDLEESGEIKDMLLECFININYIKKEEGRRFLSCLFNWNINFIKMIHGTIKNQLQGLQKSLMVYIAEIYFRAWKKAS | |
RUO | |
NCAPG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
54892 | |
LUZP5 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CAP-G2/FLJ20311/LUZP5/MTB/hCAP-G2 | |
NCAPG2 | |
Recombinant | |
wheat germ expression system |