missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LXN Partial ORF (NP_064554.2, 151 a.a. - 222 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00056925-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes the only known protein inhibitor of zinc-dependent metallocarboxypeptidases. [provided by RefSeq]
Sequence: NSTEDTWYKMVKIQTVKQVQRNDDFIELDYTILLHNIASQEIIPWQMQVLWHPQYGTKVKHNSRLPKEVQLESpecifications
NP_064554.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
33.66kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NSTEDTWYKMVKIQTVKQVQRNDDFIELDYTILLHNIASQEIIPWQMQVLWHPQYGTKVKHNSRLPKEVQLE | |
RUO | |
LXN | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
56925 | |
LXN (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ECI/TCI | |
LXN | |
Recombinant | |
wheat germ expression system |