Learn More
Abnova™ Human LY6G5B Full-length ORF (NP_067044.2, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00058496-P01.25ug
Additional Details : Weight : 0.00010kg
Description
LY6G5B belongs to a cluster of leukocyte antigen-6 (LY6) genes located in the major histocompatibility complex (MHC) class III region on chromosome 6. Members of the LY6 superfamily typically contain 70 to 80 amino acids, including 8 to 10 cysteines. Most LY6 proteins are attached to the cell surface by a glycosylphosphatidylinositol (GPI) anchor that is directly involved in signal transduction (Mallya et al., 2002 [PubMed 12079290]).[supplied by OMIM]
Sequence: MKVHMLVGVLVMVGFTVGKVPVPDIRTCHFCLVEDPSVGCISGSEKCTISSSSLCMVITIYYDVKVRFIVRGCGQYISYRCQEKRNTYFAEYWYQAQCCQYDYCNSWSSPQLQSSLPEPHDRPLALPLSDSQIQWFYQALNLSLPLPNFHAGTEPDGLDPMVTLSLNLGLSFAELRRMYLFLNSSGLLVLPQAGLLTPHPSSpecifications
NP_067044.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C6orf19/G5b | |
LY6G5B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
58496 | |
LY6G5B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKVHMLVGVLVMVGFTVGKVPVPDIRTCHFCLVEDPSVGCISGSEKCTISSSSLCMVITIYYDVKVRFIVRGCGQYISYRCQEKRNTYFAEYWYQAQCCQYDYCNSWSSPQLQSSLPEPHDRPLALPLSDSQIQWFYQALNLSLPLPNFHAGTEPDGLDPMVTLSLNLGLSFAELRRMYLFLNSSGLLVLPQAGLLTPHPS | |
RUO | |
LY6G5B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |