Läs mer
R&D Systems™ Human Lysyl Oxidase Homolog 2 DuoSet ELISA
Beskrivning
Innehåller
Capture Antibody, Detection Antibody, Recombinant Standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP)
The Human Lysyl Oxidase Homolog 2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Lysyl Oxidase Homolog 2/LOXL2.
Specifikationer
Specifikationer
| Analysintervall | 0.375 to 24 ng/mL |
| Detekterbara analyter | Lysyl Oxidase Homolog 2 |
| Analysera | Solid Phase ELISA |
| Konjugera | Biotin |
| Korsreaktivitet | <50%cross-species reactivity observed with species tested |
| Immunoassay Kit Format | 96-well Strip Plate |
| Produkttyp | ELISA Kit |
| Provtyp | Cell Culture Supernatant |
| Tillräckligt för | To run ELISAs on at least 5 x 96-well plates |
| Målarter | Human |
| Visa mer |