missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human LYZL4 Full-length ORF (NP_653235.1, 1 a.a. - 146 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_653235.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 131375 |
Molecular Weight (g/mol) | 42.8kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16187923
|
Abnova™
H00131375-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177923
|
Abnova™
H00131375-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Lysozymes (see LYZ; MIM 153450), especially C-type lysozymes, are well-recognized bacteriolytic factors widely distributed in the animal kingdom and play a mainly protective role in host defense. LYZL4 is a member of a family of lysozyme-like genes (Zhang et al., 2005 [PubMed 16014814]).[supplied by OMIM]
Sequence: MKASVVLSLLGYLVVPSGAYILGRCTVAKKLHDGGLDYFEGYSLENWVCLAYFESKFNPMAIYENTREGYTGFGLFQMRGSDWCGDHGRNRCHMSCSALLNPNLEKTIKCAKTIVKGKEGMGAWPTWSRYCQYSDTLARWLDGCKLSpecifications
NP_653235.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKASVVLSLLGYLVVPSGAYILGRCTVAKKLHDGGLDYFEGYSLENWVCLAYFESKFNPMAIYENTREGYTGFGLFQMRGSDWCGDHGRNRCHMSCSALLNPNLEKTIKCAKTIVKGKEGMGAWPTWSRYCQYSDTLARWLDGCKL | |
RUO | |
LYZL4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
131375 | |
LYZL4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
1810009N24Rik/LYC4/MGC26768 | |
LYZL4 | |
Recombinant | |
wheat germ expression system |