missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human LZIC Full-length ORF (NP_115744.2, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084328-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135843

  • 4055.00 SEK / 10µg
Estimated Shipment: 17-11-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MASRGKTETSKLKQNLEEQLDRLMQQLQDLEECREELDTDEYEETKKETLEQLSEFNDSLKKIMSGNMTLVDELSGMQLAIQAAISQAFKTPEVIRLFAKKQPGQLRTRLAEMDRDLMVGKLERDLYTQQKVEILTALRKLGEKLTADDEAFLSANAGAILSQFEKVSTDLGSGDKILALASFEVEKTKK
Specifications

Specifications

NP_115744.2
Liquid
84328
LZIC (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASRGKTETSKLKQNLEEQLDRLMQQLQDLEECREELDTDEYEETKKETLEQLSEFNDSLKKIMSGNMTLVDELSGMQLAIQAAISQAFKTPEVIRLFAKKQPGQLRTRLAEMDRDLMVGKLERDLYTQQKVEILTALRKLGEKLTADDEAFLSANAGAILSQFEKVSTDLGSGDKILALASFEVEKTKK
RUO
LZIC
Wheat Germ (in vitro)
GST
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
47.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC15436
LZIC
Yes
wheat germ expression system
Videos
SDS
Documents
Certificates
Special Offers

Special Offers