missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAGEA12 Partial ORF (NP_005358, 70 a.a. - 168 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004111-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180955

  • 5030.41 SEK / 25µg

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the MAGEA gene family. The members of this family encode proteins with 50 to 80% sequence identity to each other. The promoters and first exons of the MAGEA genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. The MAGEA genes are clustered at chromosomal location Xq28. They have been implicated in some hereditary disorders, such as dyskeratosis congenita. [provided by RefSeq]

Sequence: TLPTTINYTLWSQSDEGSSNEEQEGPSTFPDLETSFQVALSRKMAELVHFLLLKYRAREPFTKAEMLGSVIRNFQDFFPVIFSKASEYLQLVFGIEVVE
Specifications

Specifications

NP_005358
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TLPTTINYTLWSQSDEGSSNEEQEGPSTFPDLETSFQVALSRKMAELVHFLLLKYRAREPFTKAEMLGSVIRNFQDFFPVIFSKASEYLQLVFGIEVVE
RUO
MAGEA12
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4111
MAGEA12 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MAGE12
MAGEA12
Yes
Videos
SDS
Documents
Certificates