Learn More
Abnova™ Human MAGED2 Full-length ORF (NP_055414.2, 1 a.a. - 606 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010916-P01.2ug
Additional Details : Weight : 0.00010kg
Description
This gene is a member of the MAGED gene family. While the MAGEA and MAGEB genes are silent in normal tissues with the exception of testis and placenta, the MAGED genes are expressed ubiquitously. The MAGED genes are clustered on chromosome Xp11. This gene is located in Xp11.2, a hot spot for X-linked mental retardation (XLMR). Multiple alternatively spliced transcript variants have been found for this gene, however, the full length nature of some variants has not been defined. [provided by RefSeq]
Sequence: MSDTSESGAGLTRFQAEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKKVSHVADTKVNTKAQETEAAPSQAPADEPEPESAAAQSQENQDTRPKVKAKKARKVKHLDGEEDGSSDQSQASGTTGGRRVSKALMASMARRASRGPIAFWARRASRTRLAAWARRALLSLRSPKARRGKARRRAAKLQSSQEPEAPPPRDVALLQGRANDLVKYLLAKDQTKIPIKRSDMLKDIIKEYTDVYPEIIERAGYSLEKVFGIQLKEIDKNDHLYILLSTLEPTDAGILGTTKDSPKLGLLMVLLSIIFMNGNRSSEAVIWEVLRKLGLRPGIHHSLFGDVKKLITDEFVKQKYLDYARVPNSNPPEYEFFWGLRSYYETSKMKVLKFACKVQKKDPKEWAAQYREAMEADLKAAAEAAAEAKARAEIRARMGIGLGSENAAGPCNWDEADIGPWAKARIQAGAEAKAKAQESGSASTGASTSTNNSASASASTSGGFSAGASLTATLTFGLFAGLGGAGASTSGSSGACGFSYKSpecifications
NP_055414.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
91.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
11B6/BCG1/HCA10/JCL-1/MAGE-D2/MAGED/MGC8386 | |
MAGED2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10916 | |
MAGED2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSDTSESGAGLTRFQAEASEKDSSSMMQTLLTVTQNVEVPETPKASKALEVSEDVKVSKASGVSKATEVSKTPEAREAPATQASSTTQLTDTQVLAAENKSLAADTKKQNADPQAVTMPATETKKVSHVADTKVNTKAQETEAAPSQAPADEPEPESAAAQSQENQDTRPKVKAKKARKVKHLDGEEDGSSDQSQASGTTGGRRVSKALMASMARRASRGPIAFWARRASRTRLAAWARRALLSLRSPKARRGKARRRAAKLQSSQEPEAPPPRDVALLQGRANDLVKYLLAKDQTKIPIKRSDMLKDIIKEYTDVYPEIIERAGYSLEKVFGIQLKEIDKNDHLYILLSTLEPTDAGILGTTKDSPKLGLLMVLLSIIFMNGNRSSEAVIWEVLRKLGLRPGIHHSLFGDVKKLITDEFVKQKYLDYARVPNSNPPEYEFFWGLRSYYETSKMKVLKFACKVQKKDPKEWAAQYREAMEADLKAAAEAAAEAKARAEIRARMGIGLGSENAAGPCNWDEADIGPWAKARIQAGAEAKAKAQESGSASTGASTSTNNSASASASTSGGFSAGASLTATLTFGLFAGLGGAGASTSGSSGACGFSYK | |
RUO | |
MAGED2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |