missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAGI1 Partial ORF (NP_004733, 761 a.a. - 859 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004733 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9223 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16120206
|
Abnova™
H00009223-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16130206
|
Abnova™
H00009223-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]
Sequence: SHSTQVLPEFPPAEAQAPDQTDSSGQKKPDPFKIWAQSRSMYENRPMSPSPASGLSKGEREREINSTNFGECPIPDYQEQDIFLWRKETGFGFRILGGNSpecifications
NP_004733 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIP3/BAIAP1/BAP1/MAGI-1/TNRC19/WWP3 | |
MAGI1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9223 | |
MAGI1 (Human) Recombinant Protein (Q01) | |
SHSTQVLPEFPPAEAQAPDQTDSSGQKKPDPFKIWAQSRSMYENRPMSPSPASGLSKGEREREINSTNFGECPIPDYQEQDIFLWRKETGFGFRILGGN | |
RUO | |
MAGI1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |