missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAGI1 Partial ORF (NP_004733, 761 a.a. - 859 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009223-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16120206

  • 4515.00 SEK / 10µg
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the membrane-associated guanylate kinase homologue (MAGUK) family. MAGUK proteins participate in the assembly of multiprotein complexes on the inner surface of the plasma membrane at regions of cell-cell contact. The product of this gene may play a role as scaffolding protein at cell-cell junctions. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: SHSTQVLPEFPPAEAQAPDQTDSSGQKKPDPFKIWAQSRSMYENRPMSPSPASGLSKGEREREINSTNFGECPIPDYQEQDIFLWRKETGFGFRILGGN
Specifications

Specifications

NP_004733
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SHSTQVLPEFPPAEAQAPDQTDSSGQKKPDPFKIWAQSRSMYENRPMSPSPASGLSKGEREREINSTNFGECPIPDYQEQDIFLWRKETGFGFRILGGN
RUO
MAGI1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9223
MAGI1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AIP3/BAIAP1/BAP1/MAGI-1/TNRC19/WWP3
MAGI1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers