missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAN1A1 Full-length ORF (AAH65827.1, 1 a.a. - 367 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH65827.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4121 |
Molecular Weight (g/mol) | 66.4kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16137601
|
Abnova™
H00004121-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16127601
|
Abnova™
H00004121-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 23-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a class I mammalian Golgi 1,2-mannosidase which is a type II transmembrane protein. This protein catalyzes the removal of 3 distinct mannose residues from peptide-bound Man(9)-GlcNAc(2) oligosaccharides and belongs to family 47 of glycosyl hydrolases. [provided by RefSeq]
Sequence: MPVGGLLPLFSSPAGGVLGGGLGGGGGRKGSGPAALRLTEKFVLLLVFSAFITLCFGAIFFLPDSSKLLSGVLFHSSPALQPAADHKPGPGARAEDAAEGRARRREEGAPGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKIKEEIEARKGQANCPGSSSSCEVEIQMMKHAWNNYKGYAWGLNELKPISKGGHSSSLFGNIKGATIVDALDTLFIMEMKHEFEEAKSWVEENLDFNVNAEISVFEVNIRFVGGLLSAYYLSGEEIFRKKAVELGVKLLPAFHTPSGIPWALLNMKRWMCNIPRAMEATISpecifications
AAH65827.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.4kDa | |
Glutathione Sepharose 4 Fast Flow | |
MPVGGLLPLFSSPAGGVLGGGLGGGGGRKGSGPAALRLTEKFVLLLVFSAFITLCFGAIFFLPDSSKLLSGVLFHSSPALQPAADHKPGPGARAEDAAEGRARRREEGAPGDPEAALEDNLARIRENHERALREAKETLQKLPEEIQRDILLEKKKVAQDQLRDKAPFRGLPPVDFVPPIGVESREPADAAIREKRAKIKEEIEARKGQANCPGSSSSCEVEIQMMKHAWNNYKGYAWGLNELKPISKGGHSSSLFGNIKGATIVDALDTLFIMEMKHEFEEAKSWVEENLDFNVNAEISVFEVNIRFVGGLLSAYYLSGEEIFRKKAVELGVKLLPAFHTPSGIPWALLNMKRWMCNIPRAMEATI | |
RUO | |
MAN1A1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
4121 | |
MAN1A1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HUMM3/HUMM9/MAN9 | |
MAN1A1 | |
Recombinant | |
wheat germ expression system |