missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAP2K5 Partial ORF (AAH08838, 1 a.a. - 120 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

4515.00 SEK - 6845.00 SEK

Specifications

Accession Number AAH08838
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 5607
Molecular Weight (g/mol) 38.83kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16163445
View Documents Promotion Details
Abnova™
H00005607-Q01.25UG
25 ug
6845.00 SEK
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16153445
View Documents Promotion Details
Abnova™
H00005607-Q01.10UG
10 ug
4515.00 SEK
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 17-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a dual specificity protein kinase that belongs to the MAP kinase kinase family. This kinase specifically interacts with and activates MAPK7/ERK5. This kinase itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by this kinase is involved in growth factor stimulated cell proliferation and muscle cell differentiation. Four alternatively spliced transcript variants of this gene encoding distinct isoforms have been described. [provided by RefSeq]

Sequence: MLWLALGPFPAMENQVLVIRIKIPNSGAVDWTVHSGPQLLFRDVLDVIGQVLPEATTTAFEYEDEDGDRITVRSDEEMKAMLSYYYSTVMEQQVNGQLIEPLQIFPRACKPPGERNIHGL
Specifications

Specifications

AAH08838
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.83kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsT17454/MAPKK5/MEK5/PRKMK5
MAP2K5
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
5607
MAP2K5 (Human) Recombinant Protein (Q01)
MLWLALGPFPAMENQVLVIRIKIPNSGAVDWTVHSGPQLLFRDVLDVIGQVLPEATTTAFEYEDEDGDRITVRSDEEMKAMLSYYYSTVMEQQVNGQLIEPLQIFPRACKPPGERNIHGL
RUO
MAP2K5
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications