Learn More
Abnova™ Human MAP3K8 Partial ORF (NP_005195, 368 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00001326-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene was identified by its oncogenic transforming activity in cells. The encoded protein is a member of the serine/threonine protein kinase family. This kinase can activate both the MAP kinase and JNK kinase pathways. This kinase was shown to activate IkappaB kinases, and thus induce the nuclear production of NF-kappaB. This kinase was also found to promote the production of TNF-alpha and IL-2 during T lymphocyte activation. Studies of a similar gene in rat suggested the direct involvement of this kinase in the proteolysis of NF-kappaB1,p105 (NFKB1). This gene may also utilize a downstream in-frame translation start codon, and thus produce an isoform containing a shorter N-terminus. The shorter isoform has been shown to display weaker transforming activity. [provided by RefSeq]
Sequence: SLERNPNHRPRAADLLKHEALNPPREDQPRCQSLDSALLERKRLLSRKELELPENIADSSCTGSTEESEMLKRQRSLYIDLGALAGYFNLVRGPPTLEYGSpecifications
NP_005195 | |
Liquid | |
1326 | |
MAP3K8 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
COT/EST/ESTF/FLJ10486/TPL2/Tpl-2/c-COT | |
MAP3K8 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SLERNPNHRPRAADLLKHEALNPPREDQPRCQSLDSALLERKRLLSRKELELPENIADSSCTGSTEESEMLKRQRSLYIDLGALAGYFNLVRGPPTLEYG | |
RUO | |
MAP3K8 | |
Wheat Germ (in vitro) | |
GST |