missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAP4K2 Full-length ORF (NP_004570.2, 1 a.a. - 820 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_004570.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 5871 |
Molecular Weight (g/mol) | 118kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16129981
|
Abnova™
H00005871-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16139981
|
Abnova™
H00005871-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. Although this kinase is found in many tissues, its expression in lymphoid follicles is restricted to the cells of germinal centre, where it may participate in B-cell differentiation. This kinase can be activated by TNF-alpha, and has been shown to specifically activate MAP kinases. This kinase is also found to interact with TNF receptor-associated factor 2 (TRAF2), which is involved in the activation of MAP3K1/MEKK1. [provided by RefSeq]
Sequence: MALLRDVSLQDPRDRFELLQRVGAGTYGDVYKARDTVTSELAAVKIVKLDPGDDISSLQQEITILRECRHPNVVAYIGSYLRNDRLWICMEFCGGGSLQEIYHATGPLEERQIAYVCREALKGLHHLHSQGKIHRDIKGANLLLTLQGDVKLADFGVSGELTASVAKRRSFIGTPYWMAPEVAAVERKGGYNELCDVWALGITAIELGELQPPLFHLHPMRALMLMSKSSFQPPKLRDKTRWTQNFHHFLKLALTKNPKKRPTAEKLLQHPFTTQQLPRALLTQLLDKASDPHLGTPSPEDCELETYDMFPDTIHSRGQHGPAERTPSEIQFHQVKFGAPRRKETDPLNEPWEEEWTLLGKEELSGSLLQSVQEALEERSLTIRSASEFQELDSPDDTMGTIKRAPFLGPLPTDPPAEEPLSSPPGTLPPPPSGPNSSPLLPTAWATMKQREDPERSSCHGLPPTPKVHMGACFSKVFNGCPLRIHAAVTWIHPVTRDQFLVVGAEEGIYTLNLHELHEDTLEKLISHRCSWLYCVNNVLLSLSGKSTHIWAHDLPGLFEQRRLQQQVPLSIPTNRLTQRIIPRRFALSTKIPDTKGCLQCRVVRNPYTGATFLLAALPTSLLLLQWYEPLQKFLLLKNFSSPLPSPAGMLEPLVLDGKELPQVCVGAEGPEGPGCRVLFHVLPLEAGLTPDILIPPEGIPGSAQQVIQVDRDTILVSFERCVRIVNMQGEPTATLAPELTFDFPIETVVCLQDSVLAFWSHGMQGRSLDTNEVTQEITDETRIFRVLGAHRDIILESIPTDNPEAHSNLYILTGHQSTYSpecifications
NP_004570.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
118kDa | |
Glutathione Sepharose 4 Fast Flow | |
MALLRDVSLQDPRDRFELLQRVGAGTYGDVYKARDTVTSELAAVKIVKLDPGDDISSLQQEITILRECRHPNVVAYIGSYLRNDRLWICMEFCGGGSLQEIYHATGPLEERQIAYVCREALKGLHHLHSQGKIHRDIKGANLLLTLQGDVKLADFGVSGELTASVAKRRSFIGTPYWMAPEVAAVERKGGYNELCDVWALGITAIELGELQPPLFHLHPMRALMLMSKSSFQPPKLRDKTRWTQNFHHFLKLALTKNPKKRPTAEKLLQHPFTTQQLPRALLTQLLDKASDPHLGTPSPEDCELETYDMFPDTIHSRGQHGPAERTPSEIQFHQVKFGAPRRKETDPLNEPWEEEWTLLGKEELSGSLLQSVQEALEERSLTIRSASEFQELDSPDDTMGTIKRAPFLGPLPTDPPAEEPLSSPPGTLPPPPSGPNSSPLLPTAWATMKQREDPERSSCHGLPPTPKVHMGACFSKVFNGCPLRIHAAVTWIHPVTRDQFLVVGAEEGIYTLNLHELHEDTLEKLISHRCSWLYCVNNVLLSLSGKSTHIWAHDLPGLFEQRRLQQQVPLSIPTNRLTQRIIPRRFALSTKIPDTKGCLQCRVVRNPYTGATFLLAALPTSLLLLQWYEPLQKFLLLKNFSSPLPSPAGMLEPLVLDGKELPQVCVGAEGPEGPGCRVLFHVLPLEAGLTPDILIPPEGIPGSAQQVIQVDRDTILVSFERCVRIVNMQGEPTATLAPELTFDFPIETVVCLQDSVLAFWSHGMQGRSLDTNEVTQEITDETRIFRVLGAHRDIILESIPTDNPEAHSNLYILTGHQSTY | |
RUO | |
MAP4K2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
5871 | |
MAP4K2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BL44/GCK/RAB8IP | |
MAP4K2 | |
Recombinant | |
wheat germ expression system |