Learn More
Abnova™ Human MAPK3 (NP_002737.1, 1 a.a. - 379 a.a.) Full length Recombinant Protein with GST-tag
Used for Func, SDS-PAGE
Brand: Abnova™ P5772.5ug
Additional Details : Weight : 0.02000kg
Description
The protein encoded by this gene is a member of the MAP kinase family. MAP kinases, also known as extracellular signal-regulated kinases (ERKs), act in a signaling cascade that regulates various cellular processes such as proliferation, differentiation, and cell cycle progression in response to a variety of extracellular signals. This kinase is activated by upstream kinases, resulting in its translocation to the nucleus where it phosphorylates nuclear targets. Alternatively spliced transcript variants encoding different protein isoforms have been described. [provided by RefSeq]
Specifications
NP_002737.1 | |
Liquid | |
70kDa | |
Escherichia coli expression system | |
SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ERK1/HS44KDAP/HUMKER1A/MGC20180/P44ERK1/P44MAPK/PRKM3 | |
MAPK3 | |
E. coli | |
GST | |
Liquid |
Functional Study, SDS-PAGE | |
5595 | |
MAPK3 (Human) Recombinant Protein | |
Glutathione sepharose chromatography | |
5 ug | |
RUO | |
MAPK3 | |
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg (or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip™3000. Substrate : Modified Erktide. ATP: 100 µM. |
|
Recombinant | |
Escherichia coli expression system | |
90% by SDS-PAGE/CBB staining |