Learn More
Abnova™ Human MARS Partial ORF (NP_004981, 801 a.a. - 899 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004141-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. The protein encoded by this gene belongs to the class I family of tRNA synthetases. [provided by RefSeq]
Sequence: LFQKLENDQIESLRQRFGGGQAKTSPKPAVVETVTTAKPQQIQALMDEVTKQGNIVRELKAQKADKNEVAAEVAKLLDLKKQLAVAEGKPPEAPKGKKKSpecifications
NP_004981 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LFQKLENDQIESLRQRFGGGQAKTSPKPAVVETVTTAKPQQIQALMDEVTKQGNIVRELKAQKADKNEVAAEVAKLLDLKKQLAVAEGKPPEAPKGKKK | |
RUO | |
MARS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4141 | |
MARS (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ35667/METRS/MTRNS | |
MARS | |
Recombinant | |
wheat germ expression system |