missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MAS1L Partial ORF (NP_443199.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00116511-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MVWGKICWFSQRAGWTVFAESQISLSCSLCLHSGDQEAQNPNLVSQLCGVFLQNETNETIHMQMSMAVGQQALPLNIIAPKAVLVSLCGVLLNGTVFWLLSpecifications
NP_443199.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
MVWGKICWFSQRAGWTVFAESQISLSCSLCLHSGDQEAQNPNLVSQLCGVFLQNETNETIHMQMSMAVGQQALPLNIIAPKAVLVSLCGVLLNGTVFWLL | |
RUO | |
MAS1L | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
116511 | |
MAS1L (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAS-L/MGC119987/MRG/dJ994E9.2 | |
MAS1L | |
Recombinant | |
wheat germ expression system |