missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MAST1 Partial ORF (AAH27985, 203 a.a. - 311 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00022983-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16136822

  • 3343.96 SEK / 10µg

Estimated Shipment: 03-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: RFSKVYSSMEQLSQHEPKTPVAAAGSSKREPSTKGPEEKVAGKREGLGGLTLREKTWRGGSPEIKRFSASEASFLEGEASPPLGARRRFSALLEPSRFSAPQEDEDEAR
Specifications

Specifications

AAH27985
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.62kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RFSKVYSSMEQLSQHEPKTPVAAAGSSKREPSTKGPEEKVAGKREGLGGLTLREKTWRGGSPEIKRFSASEASFLEGEASPPLGARRRFSALLEPSRFSAPQEDEDEAR
RUO
MAST1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
22983
MAST1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0973/SAST/SAST170
MAST1
Yes
Videos
SDS
Documents
Certificates