Learn More
Abnova™ Human MBD3L1 Partial ORF (NP_660209.1, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00085509-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that is related to methyl-CpG-binding proteins but lacks the methyl-CpG binding domain. The protein is localized to discrete areas in the nucleus, and expression appears to be restricted to round spermatids, suggesting that the protein plays a role in the postmeiotic stages of male germ cell development. [provided by RefSeq]
Sequence: MAKSSQRKQRDCVNQCKSKPGLSTSIPLRMSSYTFKRPVTRITPHPGNEVRYHQWEESLEKPQQVCWQRRLQGLQAYSSAGELSSTLDLANTLQKLVPSYTGGSLLEDLSpecifications
NP_660209.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAKSSQRKQRDCVNQCKSKPGLSTSIPLRMSSYTFKRPVTRITPHPGNEVRYHQWEESLEKPQQVCWQRRLQGLQAYSSAGELSSTLDLANTLQKLVPSYTGGSLLEDL | |
RUO | |
MBD3L1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
85509 | |
MBD3L1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MBD3L/MGC138263/MGC138269 | |
MBD3L1 | |
Recombinant | |
wheat germ expression system |