missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MED12 Partial ORF (NP_005111, 1829 a.a. - 1941 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00009968-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16191536

  • 4515.00 SEK / 10µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The initiation of transcription is controlled in part by a large protein assembly known as the preinitiation complex. A component of this preinitiation complex is a 1.2 MDa protein aggregate called Mediator. This Mediator component binds with a CDK8 subcomplex which contains the protein encoded by this gene, mediator complex subunit 12 (MED12), along with MED13, CDK8 kinase, and cyclin C. The CDK8 subcomplex modulates Mediator-polymerase II interactions and thereby regulates transcription initiation and reinitation rates. The MED12 protein is essential for activating CDK8 kinase. Defects in this gene cause X-linked Opitz-Kaveggia syndrome, also known as FG syndrome, and Lujan-Fryns syndrome. [provided by RefSeq]

Sequence: QPATKTEDYGMGPGRSGPYGVTVPPDLLHHPNPGSITHLNYRQGSIGLYTQNQPLPAGGPRVDPYRPVRLPMQKLPTRPTYPGVLPTTMTGVMGLEPSSYKTSVYRQQQPAVP
Specifications

Specifications

NP_005111
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
38.17kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QPATKTEDYGMGPGRSGPYGVTVPPDLLHHPNPGSITHLNYRQGSIGLYTQNQPLPAGGPRVDPYRPVRLPMQKLPTRPTYPGVLPTTMTGVMGLEPSSYKTSVYRQQQPAVP
RUO
MED12
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
9968
MED12 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CAGH45/FGS1/HOPA/KIAA0192/OKS/OPA1/TNRC11/TRAP230
MED12
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers