Learn More
Abnova™ Human MED29 Full-length ORF (NP_060062.1, 1 a.a. - 221 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055588-P01.25ug
Additional Details : Weight : 0.00010kg
Description
MED29 is a subunit of the Mediator complex, a multiprotein coactivator of RNA transcription that interacts with DNA-bound transcriptional activators, RNA polymerase II (see MIM 180660), and general initiation factors (Sato et al., 2003 [PubMed 14576168]).[supplied by OMIM]
Sequence: MLKSNGERRSRNALPAVYARKMAASQQQASAASSAAGVSGPSSAGGPGPQQQPQPPAQLVGPAQSGLLQQQQQDFDPVQRYKMLIPQLKESLQTLMKVAAQNLIQNTNIDNGQKSSDGPIQRFDKCLEEFYALCDQLELCLRLAHECLSQSCDSAKHSPTLVPTATKPDAVQPDSLPYPQYLAVIKAQISCAKDIHTALLDCANKVTGKTPAPPAGPGGTLSpecifications
NP_060062.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434H247/IXL | |
MED29 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
55588 | |
MED29 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MLKSNGERRSRNALPAVYARKMAASQQQASAASSAAGVSGPSSAGGPGPQQQPQPPAQLVGPAQSGLLQQQQQDFDPVQRYKMLIPQLKESLQTLMKVAAQNLIQNTNIDNGQKSSDGPIQRFDKCLEEFYALCDQLELCLRLAHECLSQSCDSAKHSPTLVPTATKPDAVQPDSLPYPQYLAVIKAQISCAKDIHTALLDCANKVTGKTPAPPAGPGGTL | |
RUO | |
MED29 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |