Läs mer
STEMCELL Technologies Human Mesenchymal Stem Cell qPCR Array
Beskrivning
- The Human Mesenchymal Stem Cell Quantitative Polymerase Chain Reaction (qPCR) Array is designed for characterization of human mesenchymal stem and progenitor cells (MSCs) and their differentiated chondrogenic, adipogenic, or osteogenic progeny
- MSCs can be expanded in vitro to generate mesenchymal stromal cell cultures, which, under appropriate conditions, can differentiate into adipocytes, chondrocytes, and osteoblasts
- This qPCR array provides the gene expression profile of MSCs and their trilineage derivatives following in vitro differentiation
Specifikationer
Specifikationer
| Beskrivning | Adipocytes; Chondrocytes; Mesenchymal Stem and Progenitor Cells; Osteoblasts |
| Formatera | 96-well Plate Type 3 |
| För användning med (applikation) | Real Time PCR (qPCR) |