Läs mer
Human MFAP3 Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB113251V-100UG
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MFAP3 Monoclonal antibody specifically detects MFAP3 in Human samples. It is validated for Flow Cytometry
Specifikationer
| MFAP3 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3 | |
| Human embryonic kidney cell, HEK293-derived human MFAP3L, Met1-Met149, Accession # O75121 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 1059143 | |
| Flow Cytometry | |
| O75121 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4238 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
| IgG2b |