missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MFAP4 Full-length ORF (NP_002395.1, 1 a.a. - 255 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_002395.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4239 |
Molecular Weight (g/mol) | 55kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16187771
|
Abnova™
H00004239-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16177771
|
Abnova™
H00004239-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a protein with similarity to a bovine microfibril-associated protein. The protein has binding specificities for both collagen and carbohydrate. It is thought to be an extracellular matrix protein which is involved in cell adhesion or intercellular interactions. The gene is located within the Smith-Magenis syndrome region. [provided by RefSeq]
Sequence: MKALLALPLLLLLSTPPCAPQVSGIRGDALERFCLQQPLDCDDIYAQGYQSDGVYLIYPSGPSVPVPVFCDMTTEGGKWTVFQKRFNGSVSFFRGWNDYKLGFGRADGEYWLGLQNMHLLTLKQKYELRVDLEDFENNTAYAKYADFSISPNAVSAEEDGYTLFVAGFEDGGAGDSLSYHSGQKFSTFDRDQDLFVQNCAALSSGAFWFRSCHFANLNGFYLGGSHLSYANGINWAQWKGFYYSLKRTEMKIRRASpecifications
NP_002395.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55kDa | |
Glutathione Sepharose 4 Fast Flow | |
MKALLALPLLLLLSTPPCAPQVSGIRGDALERFCLQQPLDCDDIYAQGYQSDGVYLIYPSGPSVPVPVFCDMTTEGGKWTVFQKRFNGSVSFFRGWNDYKLGFGRADGEYWLGLQNMHLLTLKQKYELRVDLEDFENNTAYAKYADFSISPNAVSAEEDGYTLFVAGFEDGGAGDSLSYHSGQKFSTFDRDQDLFVQNCAALSSGAFWFRSCHFANLNGFYLGGSHLSYANGINWAQWKGFYYSLKRTEMKIRRA | |
RUO | |
MFAP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
4239 | |
MFAP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MFAP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |