missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MGC40579 Full-length ORF (AAH32470.1, 1 a.a. - 301 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00256356-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MSGLLTDPEQRAQEPRYPGFVLGLDVGSSVIRCHVYDRAARVCGSSVQKVENLYPQIGWVEIDPDVLWIQFVAVIKEAVKAAGIQMNQIVGLGISTQRATFITWNKKTGNHFHNFISWQDLRAVELVKSWNNSLLMKIFHSSCRVLHFFTRSKRLFTASLFTFTTQQTSLRLVWILQNLTEVQKAVEEENCCFGTIDTWLLYKLTKGSVYATDFSNASTTGLFDPYKMCWSGMITSLISIPLSLLPPVRDTSHNFGSVDEEIFGVPIPIVALLKVPGYDQNICYIFGKGTIEPVFYHGSTLSpecifications
AAH32470.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ33582/FLJ45739/MGC40579 | |
GK5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
256356 | |
MGC40579 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSGLLTDPEQRAQEPRYPGFVLGLDVGSSVIRCHVYDRAARVCGSSVQKVENLYPQIGWVEIDPDVLWIQFVAVIKEAVKAAGIQMNQIVGLGISTQRATFITWNKKTGNHFHNFISWQDLRAVELVKSWNNSLLMKIFHSSCRVLHFFTRSKRLFTASLFTFTTQQTSLRLVWILQNLTEVQKAVEEENCCFGTIDTWLLYKLTKGSVYATDFSNASTTGLFDPYKMCWSGMITSLISIPLSLLPPVRDTSHNFGSVDEEIFGVPIPIVALLKVPGYDQNICYIFGKGTIEPVFYHGSTL | |
RUO | |
GK5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |