Learn More
Abnova™ Human MIPEP Partial ORF (NP_005923, 611 a.a. - 713 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00004285-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The product of this gene performs the final step in processing a specific class of nuclear-encoded proteins targeted to the mitochondrial matrix or inner membrane. This protein is primarily involved in the maturation of oxidative phosphorylation (OXPHOS)-related proteins. This gene may contribute to the functional effects of frataxin deficiency and the clinical manifestations of Friedreich ataxia. [provided by RefSeq]
Sequence: PYVPNTAWQLRFSHLVGYGARYYSYLMSRAVASMVWKECFLQDPFNRAAGERYRREMLAHGGGREPMLMVEGMLQKCPSVDDFVSALVSDLDLDFETFLMDSESpecifications
NP_005923 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PYVPNTAWQLRFSHLVGYGARYYSYLMSRAVASMVWKECFLQDPFNRAAGERYRREMLAHGGGREPMLMVEGMLQKCPSVDDFVSALVSDLDLDFETFLMDSE | |
RUO | |
MIPEP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
4285 | |
MIPEP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HMIP/MIP | |
MIPEP | |
Recombinant | |
wheat germ expression system |