missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MKLN1 Partial ORF (NP_037387, 633 a.a. - 735 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004289-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16121275

  • 4515.00 SEK / 10µg
Estimated Shipment: 23-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Muskelin is an intracellular protein that acts as a mediator of cell spreading and cytoskeletal responses to the extracellular matrix component thrombospondin I (MIM 188060) (Adams et al., 1998 [PubMed 9724633]).[supplied by OMIM]

Sequence: RHCKYLIRKHRFEEKAQVDPLSALKYLQNDLYITVDHSDPEETKEFQLLASALFKSGSDFTALGFSDVDHTYAQRTQLFDTLVNFFPDSMTPPKGNLVDLITL
Specifications

Specifications

NP_037387
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.07kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RHCKYLIRKHRFEEKAQVDPLSALKYLQNDLYITVDHSDPEETKEFQLLASALFKSGSDFTALGFSDVDHTYAQRTQLFDTLVNFFPDSMTPPKGNLVDLITL
RUO
MKLN1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4289
MKLN1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11162/TWA2
MKLN1
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers