missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MLLT10 Partial ORF (NP_001009569, 695 a.a. - 793 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008028-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16118155

  • 6845.00 SEK / 25µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: QIRYDQPGNSSLENLPPVAASIEQLLERQWSEGQQFLLEQGTPSDILGMLKSLHQLQVENRRLEEQIKNLTAKKERLQLLNAQLSVPFPTITANPSPSH
Specifications

Specifications

NP_001009569
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QIRYDQPGNSSLENLPPVAASIEQLLERQWSEGQQFLLEQGTPSDILGMLKSLHQLQVENRRLEEQIKNLTAKKERLQLLNAQLSVPFPTITANPSPSH
RUO
MLLT10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8028
MLLT10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AF10/DKFZp686E10210/MGC75086
MLLT10
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers