Learn More
Abnova™ Human MMAB Partial ORF (NP_443077.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00326625-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a protein that catalyzes the final step in the conversion of vitamin B(12) into adenosylcobalamin (AdoCbl), a vitamin B12-containing coenzyme for methylmalonyl-CoA mutase. Mutations in the gene are the cause of vitamin B12-dependent methylmalonic aciduria linked to the cblB complementation group. [provided by RefSeq]
Sequence: MAVCGLGSRLGLGSRLGLRGCFGAARLLYPRFQSRGPQGVEDGDRPQPSSKTPRIPKIYTKTGDKGFSSTFTGERRPKDDQVFEAVGTTDELSSAIGFALSpecifications
NP_443077.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAVCGLGSRLGLGSRLGLRGCFGAARLLYPRFQSRGPQGVEDGDRPQPSSKTPRIPKIYTKTGDKGFSSTFTGERRPKDDQVFEAVGTTDELSSAIGFAL | |
RUO | |
MMAB | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
326625 | |
MMAB (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATR/MGC20496 | |
MMAB | |
Recombinant | |
wheat germ expression system |