missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MMEL1 Full-length ORF (AAI01031.1, 1 a.a. - 779 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079258-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16184173

  • 5030.41 SEK / 25µg

Estimated Shipment: 07-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. This protein is a type II transmembrane protein and is thought to be expressed as a secreted protein. This gene is expressed mainly in testis with weak expression in the brain, kidney, and heart. [provided by RefSeq]

Sequence: MGKSEGPVGMVESAGRAGQKRPGFLEGGLLLLLLLVTAALVALGVLYADRRGKQLPRLASRLCFLQEERTFVKRKPRGIPEAQEVSEVCTTPGCVIAAARILQNMDPTTEPCDDFYQFACGGWLRRHVIPETNSRYSIFDVLRDELEVILKAVLENSTAKDRPAVEKARTLYRSCMNQSVIEKRGSQPLLDILEVVGGWPVAMDRWNETVGLEWELERQLALMNSQFNRRVLIDLFIWNDDQNSSRHIIYIDQPTLGMPSREYYFNGGSNRKVREAYLQFMVSVATLLREDANLPRDSCLVQEDMVQVLELETQLAKATVPQEERHDVIALYHRMGLEELQSQFGLKGFNWTLFIQTVLSSVKIKLLPDEEVVVYGIPYLQNLENIIDTYSARTIQNYLVWRLVLDRIGSLSQRFKDTRVNYRKALFGTMVEEVRWRECVGYVNSNMENAVGSLYVREAFPGDSKSMVRELIDKVRTVFVETLDELGWIDEESKKKAQEKAMSIREQIGHPDYILEETNRRLDEEYSNLNFSEDLYFENSLQNLKVGAQRSLRKLREKVDPNLWIIGAAVVNAFYSPNRNQIVFPAGILQPPFFSKEQPQALNFGGIGMVIGHEITHGFDDNGRNFDKNGNMMDWWSNFSTQHFREQSECMIYQYGNYSWDLADEQNVNGFNTLGENIADNGGVRQAYKAYLKWMAEGGKDQQLPGLDLTHEQLFFINYAQVWCGSYRPEFAIQSIKTDVHSPLKYRVLGSLQNLAAFADTFHCARGTPMHPKERCRVW
Specifications

Specifications

AAI01031.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
115.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC119454/MGC119455/MGC119456/MMEL2/NEP2/NEPII/NL1/NL2/SEP
MMEL1
Yes
wheat germ expression system
Liquid
79258
MMEL1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGKSEGPVGMVESAGRAGQKRPGFLEGGLLLLLLLVTAALVALGVLYADRRGKQLPRLASRLCFLQEERTFVKRKPRGIPEAQEVSEVCTTPGCVIAAARILQNMDPTTEPCDDFYQFACGGWLRRHVIPETNSRYSIFDVLRDELEVILKAVLENSTAKDRPAVEKARTLYRSCMNQSVIEKRGSQPLLDILEVVGGWPVAMDRWNETVGLEWELERQLALMNSQFNRRVLIDLFIWNDDQNSSRHIIYIDQPTLGMPSREYYFNGGSNRKVREAYLQFMVSVATLLREDANLPRDSCLVQEDMVQVLELETQLAKATVPQEERHDVIALYHRMGLEELQSQFGLKGFNWTLFIQTVLSSVKIKLLPDEEVVVYGIPYLQNLENIIDTYSARTIQNYLVWRLVLDRIGSLSQRFKDTRVNYRKALFGTMVEEVRWRECVGYVNSNMENAVGSLYVREAFPGDSKSMVRELIDKVRTVFVETLDELGWIDEESKKKAQEKAMSIREQIGHPDYILEETNRRLDEEYSNLNFSEDLYFENSLQNLKVGAQRSLRKLREKVDPNLWIIGAAVVNAFYSPNRNQIVFPAGILQPPFFSKEQPQALNFGGIGMVIGHEITHGFDDNGRNFDKNGNMMDWWSNFSTQHFREQSECMIYQYGNYSWDLADEQNVNGFNTLGENIADNGGVRQAYKAYLKWMAEGGKDQQLPGLDLTHEQLFFINYAQVWCGSYRPEFAIQSIKTDVHSPLKYRVLGSLQNLAAFADTFHCARGTPMHPKERCRVW
RUO
MMEL1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates