missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MMEL1 Full-length ORF (AAI01031.1, 1 a.a. - 779 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAI01031.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79258 |
Molecular Weight (g/mol) | 115.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16184173
|
Abnova™
H00079258-P01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16174173
|
Abnova™
H00079258-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The protein encoded by this gene is a member of the neutral endopeptidase (NEP) or membrane metallo-endopeptidase (MME) family. Family members play important roles in pain perception, arterial pressure regulation, phosphate metabolism and homeostasis. This protein is a type II transmembrane protein and is thought to be expressed as a secreted protein. This gene is expressed mainly in testis with weak expression in the brain, kidney, and heart. [provided by RefSeq]
Sequence: MGKSEGPVGMVESAGRAGQKRPGFLEGGLLLLLLLVTAALVALGVLYADRRGKQLPRLASRLCFLQEERTFVKRKPRGIPEAQEVSEVCTTPGCVIAAARILQNMDPTTEPCDDFYQFACGGWLRRHVIPETNSRYSIFDVLRDELEVILKAVLENSTAKDRPAVEKARTLYRSCMNQSVIEKRGSQPLLDILEVVGGWPVAMDRWNETVGLEWELERQLALMNSQFNRRVLIDLFIWNDDQNSSRHIIYIDQPTLGMPSREYYFNGGSNRKVREAYLQFMVSVATLLREDANLPRDSCLVQEDMVQVLELETQLAKATVPQEERHDVIALYHRMGLEELQSQFGLKGFNWTLFIQTVLSSVKIKLLPDEEVVVYGIPYLQNLENIIDTYSARTIQNYLVWRLVLDRIGSLSQRFKDTRVNYRKALFGTMVEEVRWRECVGYVNSNMENAVGSLYVREAFPGDSKSMVRELIDKVRTVFVETLDELGWIDEESKKKAQEKAMSIREQIGHPDYILEETNRRLDEEYSNLNFSEDLYFENSLQNLKVGAQRSLRKLREKVDPNLWIIGAAVVNAFYSPNRNQIVFPAGILQPPFFSKEQPQALNFGGIGMVIGHEITHGFDDNGRNFDKNGNMMDWWSNFSTQHFREQSECMIYQYGNYSWDLADEQNVNGFNTLGENIADNGGVRQAYKAYLKWMAEGGKDQQLPGLDLTHEQLFFINYAQVWCGSYRPEFAIQSIKTDVHSPLKYRVLGSLQNLAAFADTFHCARGTPMHPKERCRVWSpecifications
AAI01031.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
115.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGKSEGPVGMVESAGRAGQKRPGFLEGGLLLLLLLVTAALVALGVLYADRRGKQLPRLASRLCFLQEERTFVKRKPRGIPEAQEVSEVCTTPGCVIAAARILQNMDPTTEPCDDFYQFACGGWLRRHVIPETNSRYSIFDVLRDELEVILKAVLENSTAKDRPAVEKARTLYRSCMNQSVIEKRGSQPLLDILEVVGGWPVAMDRWNETVGLEWELERQLALMNSQFNRRVLIDLFIWNDDQNSSRHIIYIDQPTLGMPSREYYFNGGSNRKVREAYLQFMVSVATLLREDANLPRDSCLVQEDMVQVLELETQLAKATVPQEERHDVIALYHRMGLEELQSQFGLKGFNWTLFIQTVLSSVKIKLLPDEEVVVYGIPYLQNLENIIDTYSARTIQNYLVWRLVLDRIGSLSQRFKDTRVNYRKALFGTMVEEVRWRECVGYVNSNMENAVGSLYVREAFPGDSKSMVRELIDKVRTVFVETLDELGWIDEESKKKAQEKAMSIREQIGHPDYILEETNRRLDEEYSNLNFSEDLYFENSLQNLKVGAQRSLRKLREKVDPNLWIIGAAVVNAFYSPNRNQIVFPAGILQPPFFSKEQPQALNFGGIGMVIGHEITHGFDDNGRNFDKNGNMMDWWSNFSTQHFREQSECMIYQYGNYSWDLADEQNVNGFNTLGENIADNGGVRQAYKAYLKWMAEGGKDQQLPGLDLTHEQLFFINYAQVWCGSYRPEFAIQSIKTDVHSPLKYRVLGSLQNLAAFADTFHCARGTPMHPKERCRVW | |
RUO | |
MMEL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79258 | |
MMEL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119454/MGC119455/MGC119456/MMEL2/NEP2/NEPII/NL1/NL2/SEP | |
MMEL1 | |
Recombinant | |
wheat germ expression system |