missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MOGAT3 Full-length ORF (NP_835470.1, 1 a.a. - 341 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00346606-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16180274

  • 3343.96 SEK / 10µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Acyl-CoA:monoacylglycerol acyltransferase (MOGAT; EC 2.3.1.22) catalyzes the synthesis of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA (Cheng et al., 2003 [PubMed 12618427]).[supplied by OMIM]

Sequence: MGVATTLQPPTTSKTLQKQHLEAVGAYQYVLTFLFMGPFFSLLVFVLLFTSLWPFSVFYLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDRNYVLGAHPHGIMCTGFLCNFSTESNGFSQLFPGLRPWLAVLAGLFYLPVYRDYIMSFGLCPVSRQSLDFILSQPQLGQAVVIMVGGAHEALYSVPGEHCLTLQKRKGFVRLALRHGASLVPVYSFGENDIFRLKAFATGSWQHWCQLTFKKLMGFSPCIFWGRGLFSATSWGLLPFAVPITTVVGRPIPVPQRLHPTEEEVNHYHALYMTALEQLFEEHKESCGVPASTCLTFI
Specifications

Specifications

NP_835470.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DC7/DGAT2L7/MGAT3/MGC119203/MGC119204
MOGAT3
Yes
wheat germ expression system
Liquid
346606
MOGAT3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGVATTLQPPTTSKTLQKQHLEAVGAYQYVLTFLFMGPFFSLLVFVLLFTSLWPFSVFYLVWLYVDWDTPNQGGRRSEWIRNRAIWRQLRDYYPVKLVKTAELPPDRNYVLGAHPHGIMCTGFLCNFSTESNGFSQLFPGLRPWLAVLAGLFYLPVYRDYIMSFGLCPVSRQSLDFILSQPQLGQAVVIMVGGAHEALYSVPGEHCLTLQKRKGFVRLALRHGASLVPVYSFGENDIFRLKAFATGSWQHWCQLTFKKLMGFSPCIFWGRGLFSATSWGLLPFAVPITTVVGRPIPVPQRLHPTEEEVNHYHALYMTALEQLFEEHKESCGVPASTCLTFI
RUO
MOGAT3
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates