missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human/Mouse Phospho-STAT1 (Y701) Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF2894
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
STAT1 Polyclonal specifically detects STAT1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifikationer
| STAT1 (p Tyr701) | |
| Polyclonal | |
| Unconjugated | |
| CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 6772 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| STAT1 | |
| Phosphopeptide containing human STAT1 Y701 site | |
| 50 μg | |
| Primary | |
| Detects human STAT1 when phosphorylated at Y701 in Western blots. | |
| Human, Mouse | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering