missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human/Mouse/Rat/Chicken Oligodendrocyte Marker O4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 59 publications
Brand: R&D Systems MAB1326-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifikationer
| Oligodendrocyte Marker O4 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 1-10 ug/mL, CyTOF-ready | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4. | |
| Human, Mouse, Rat, Chicken | |
| IgM |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| O4 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bovine brain corpus callosum white matter | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering