missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human/Mouse/Rat NEDD8 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4936
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
NEDD8 Polyclonal specifically detects NEDD8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifikationer
| NEDD8 | |
| Polyclonal | |
| Unconjugated | |
| Q15843 | |
| NEDD8 | |
| E. coli-derived recombinant human NEDD8 Leu2-Gly77 Accession # Q15843 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat NEDD8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human NEDD9 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 2 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| FLJ43224, MGC104393, MGC125896, MGC125897, NEDD-8, Neddylin, Neural precursor cell expressed developmentally down-regulated protein 8, neural precursor cell expressed, developmentally down-regulated 8, Rub1, Ubiquitin-like protein Nedd8 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4738 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering