missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human/Mouse/Rat PI 3-Kinase p85 alpha Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2998
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
PI 3-Kinase p85 alpha Polyclonal specifically detects PI 3-Kinase p85 alpha in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifikationer
| PI 3-Kinase p85 alpha | |
| Polyclonal | |
| Unconjugated | |
| P27986 | |
| PIK3R1 | |
| E. coli-derived recombinant human PI 3-Kinase p85 alpha Leu328-Tyr431 Accession # P27986 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat PI 3-Kinase p85 alpha in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AGM7, GRB1, IMD36, p85, p85-ALPHA, Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha, phosphatidylinositol 3-kinase regulatory subunit alpha, phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha), phosphatidylinositol 3-kinase-associated p-85 alpha, phosphoinositide-3-kinase regulatory subunit alpha, phosphoinositide-3-kinase, regulatory subunit 1 (alpha), PI 3Kinase p85 alpha, PI3K regulatory subunit alpha, PI3-kinase regulatory subunit alpha, PI3-kinase subunit p85-alpha, PIK3R1, PtdIns-3-kinase regulatory subunit alpha, PtdIns-3-kinase regulatory subunit p85-alpha | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5295 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering