missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Human/Mouse TGF-beta RI/ALK-5 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5871R-100
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
TGF-beta RI/ALK-5 Monoclonal specifically detects TGF-beta RI/ALK-5 in Human, Mouse samples. It is validated for Western Blot.
Specifikationer
| TGF-beta RI/ALK-5 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AAT5, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, Activin receptor-like kinase 5, ACVRLK4, ALK-5, ALK-5ALK5, EC 2.7.11, EC 2.7.11.30, LDS1A, LDS2A, Serine/threonine-protein kinase receptor R4, SKR4, TbetaR-I, TGFB1R1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, TGFbetaRI, TGFBR1, TGF-bRI, TGFR-1, transforming growth factor beta receptor I, transforming growth factor, beta receptor 1, transforming growth factor, beta receptor I (activin A receptor type II-likekinase, 53kD), Transforming growth factor-beta receptor type I | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse TGF-beta RI/ALK-5, Ala21-Glu121, Accession # BAA05023 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 141231R | |
| Western Blot 1 ug/mL | |
| BAA05023 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7046 | |
| Human, Mouse | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering