missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MPHOSPH9 Full-length ORF (NP_073619.2, 1 a.a. - 1031 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00010198-P01.10ug
Additional Details : Weight : 0.00010kg
Description
Sequence: MGFFSLSSERNESVIHYPESTEPEIQQEMSTSQPDCNVDSCSVSSGYGTFCISELNLYKSKDPKEFMEHIDVPKGQYVAPAVPAESLVDGVKNENFYIQTPEECHVSLKEDVSISPGEFEHNFLGENKVSEVYSGKTNSNAITSWAQKLKQNQPKRAHVEDGGSRSKQGNEQSKKTPIEKSDFAAATHPRAFYLSKPDETPNAWMSDSGTGLTYWKLEEKDMHHSLPETLEKTFISLSSTDVSPNQSNTSNEMKLPSLKDIYYKKQRENKQLPERNLTSASNPNHPPEVLTLDPTLHMKPKQQISGIQPHGLPNALDDRISFSPDSVLEPSMSSPSDIDSFSQASNVTSQLPGFPKYPSHTKASPVDSWKNQTFQNESRTSSTFPSVYTITSNDISVNTVDEENTVMVASASVSQSQLPGTANSVPECISLTSLEDPVILSKIRQNLKEKHARHIADLRAYYESEINSLKQKLEAKEISGVEDWKITNQILVDRCGQLDSALHEATSRVRTLENKNNLLEIEVNDLRERFSAASSASKILQERIEEMRTSSKEKDNTIIRLKSRLQDLEEAFENAYKLSDDKEAQLKQENKMFQDLLGEYESLGKEHRRVKDALNTTENKLLDAYTQISDLKRMISKLEAQVKQVEHENMLSLRHNSRIHVRPSRANTLATSDVSRRKWLIPGAEYSIFTGQPLDTQDSNVDNQLEETCSLGHRSPLEKDSSPGSSSTSLLIKKQRETSDTPIMRALKELDEGKIFKNWGTQTEKEDTSNINPRQTETSVNASRSPEKCAQQRQKRLNSASQRSSSLPPSNRKSSTPTKREIMLTPVTVAYSPKRSPKENLSPGFSHLLSKNESSPIRFDILLDDLDTVPVSTLQRTNPRKQLQFLPLDDSEEKTYSEKATDNHVNHSSCPEPVPNGVKKVSVRTAWEKNKSVSYEQCKPVSVTPQGNDFEYTAKIRTLAETERFFDELTKEKDQIEAALSRMPSPGGRITLQTRLNQEALEDRLERINRELGSVRMTLKKFHVLRTSANLSpecifications
NP_073619.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
142.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp434J034/FLJ12954/MPP-9/MPP9 | |
MPHOSPH9 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
10198 | |
MPHOSPH9 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGFFSLSSERNESVIHYPESTEPEIQQEMSTSQPDCNVDSCSVSSGYGTFCISELNLYKSKDPKEFMEHIDVPKGQYVAPAVPAESLVDGVKNENFYIQTPEECHVSLKEDVSISPGEFEHNFLGENKVSEVYSGKTNSNAITSWAQKLKQNQPKRAHVEDGGSRSKQGNEQSKKTPIEKSDFAAATHPRAFYLSKPDETPNAWMSDSGTGLTYWKLEEKDMHHSLPETLEKTFISLSSTDVSPNQSNTSNEMKLPSLKDIYYKKQRENKQLPERNLTSASNPNHPPEVLTLDPTLHMKPKQQISGIQPHGLPNALDDRISFSPDSVLEPSMSSPSDIDSFSQASNVTSQLPGFPKYPSHTKASPVDSWKNQTFQNESRTSSTFPSVYTITSNDISVNTVDEENTVMVASASVSQSQLPGTANSVPECISLTSLEDPVILSKIRQNLKEKHARHIADLRAYYESEINSLKQKLEAKEISGVEDWKITNQILVDRCGQLDSALHEATSRVRTLENKNNLLEIEVNDLRERFSAASSASKILQERIEEMRTSSKEKDNTIIRLKSRLQDLEEAFENAYKLSDDKEAQLKQENKMFQDLLGEYESLGKEHRRVKDALNTTENKLLDAYTQISDLKRMISKLEAQVKQVEHENMLSLRHNSRIHVRPSRANTLATSDVSRRKWLIPGAEYSIFTGQPLDTQDSNVDNQLEETCSLGHRSPLEKDSSPGSSSTSLLIKKQRETSDTPIMRALKELDEGKIFKNWGTQTEKEDTSNINPRQTETSVNASRSPEKCAQQRQKRLNSASQRSSSLPPSNRKSSTPTKREIMLTPVTVAYSPKRSPKENLSPGFSHLLSKNESSPIRFDILLDDLDTVPVSTLQRTNPRKQLQFLPLDDSEEKTYSEKATDNHVNHSSCPEPVPNGVKKVSVRTAWEKNKSVSYEQCKPVSVTPQGNDFEYTAKIRTLAETERFFDELTKEKDQIEAALSRMPSPGGRITLQTRLNQEA | |
RUO | |
MPHOSPH9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |