missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MPP2 Partial ORF (NP_005365.3, 72 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | NP_005365.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4355 |
Molecular Weight (g/mol) | 36.08kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16141355
|
Abnova™
H00004355-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16131355
|
Abnova™
H00004355-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Palmitoylated membrane protein 2 is a member of a family of membrane-associated proteins termed MAGUKs (membrane-associated guanylate kinase homologs). MAGUKs interact with the cytoskeleton and regulate cell proliferation, signaling pathways, and intracellular junctions. Palmitoylated membrane protein 2 contains a conserved sequence, called the SH3 (src homology 3) motif, found in several other proteins that associate with the cytoskeleton and are suspected to play important roles in signal transduction. [provided by RefSeq]
Sequence: QEILRDLAHVAEQSSTAAELAHILQEPHFQSLLETHDSVASKTYETPPPSPGLDPTFSNQPVPPDAVRMVGIRKTAGEHLGVTFRVEGGELVIASpecifications
NP_005365.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.08kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686A06252/DKFZp686J2189/DKFZp761D0712/DLG2 | |
MPP2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4355 | |
MPP2 (Human) Recombinant Protein (Q01) | |
QEILRDLAHVAEQSSTAAELAHILQEPHFQSLLETHDSVASKTYETPPPSPGLDPTFSNQPVPPDAVRMVGIRKTAGEHLGVTFRVEGGELVIA | |
RUO | |
MPP2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |