Läs mer
R&D Systems™ Human MRGX1 (NP_671732) VersaClone cDNA
Produktkod. 18716728
Change view
Kvantitet:
10 μg
Produktkod. Kvantitet
18716728 10 μg
1 options
Produktkod. 18716728 Leverantör R&D Systems™ Leverantörsnummer RDC0959
  • ONLINE EXCLUSIVE PRICE
    3500.00 SEK
    3309.60 SEK / 10µg
    Save 190.40 SEK
    5% Av
 Begränsad artikel
Beräknad leverans: 11-11-2026
MAS-related GPR, member X1
The Human MRGX1 (NP_671732) VersaClone cDNA clone contains the complete ORF for MRGX1, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The MRGX1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Beställningsinfo
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifikationer
Innehåll och lagring Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Kvantitet 10 μg
For Reasearch Use Only; Not for Use in Humans