missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MRPL49 Full-length ORF (NP_004918.1, 1 a.a. - 166 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00000740-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16135857

  • 3343.96 SEK / 10µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene and the gene for the HRD1 protein use in their respective 3' UTRs some of the same genomic sequence. Pseudogenes corresponding to this gene are found on chromosomes 5q and 8p. [provided by RefSeq]

Sequence: MAATMFRATLRGWRTGVQRGCGLRLLSQTQGPPDYPRFVESVDEYQFVERLLPATRIPDPPKHEHYPTPSGWQPPRDPPPNLPYFVRRSRMHNIPVYKDITHGNRQMTVIRKVEGDIWALQKDVEDFLSPLLGKTPVTQVNEVTGTLRIKGYFDQELKAWLLEKGF
Specifications

Specifications

NP_004918.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C11orf4/L49mt/MGC10656/NOF/NOF1
MRPL49
Yes
wheat germ expression system
Liquid
740
MRPL49 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAATMFRATLRGWRTGVQRGCGLRLLSQTQGPPDYPRFVESVDEYQFVERLLPATRIPDPPKHEHYPTPSGWQPPRDPPPNLPYFVRRSRMHNIPVYKDITHGNRQMTVIRKVEGDIWALQKDVEDFLSPLLGKTPVTQVNEVTGTLRIKGYFDQELKAWLLEKGF
RUO
MRPL49
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates