Learn More
Abnova™ Human MS4A3 Full-length ORF (NP_006129.4, 1 a.a. - 214 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00000932-P02.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the membrane-spanning 4A gene family. Members of this protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. This family member likely plays a role in signal transduction and may function as a subunit associated with receptor complexes. The gene encoding this protein is localized to 11q12, among a cluster of related family members. Alternative splicing may result in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq]
Sequence: MASHEVDNAELGSASAHGTPGSEAGPEELNTSVYQPIDGSPDYQKAKLQVLGAIQILNAAMILALGVFLGSLQYPYHFQKHFFFFTFYTGYPIWGAVFFCSSGTLSVVAGIKPTRTWIQNSFGMNIASATIALVGTAFLSLNIAVNIQSLRSCHSSSESPDLCNYMGSISNGMVSLLLILTLLELCVTISTIAMWCNANCCNSREEISSPPNSVSpecifications
NP_006129.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD20L/HTM4 | |
MS4A3 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
932 | |
MS4A3 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASHEVDNAELGSASAHGTPGSEAGPEELNTSVYQPIDGSPDYQKAKLQVLGAIQILNAAMILALGVFLGSLQYPYHFQKHFFFFTFYTGYPIWGAVFFCSSGTLSVVAGIKPTRTWIQNSFGMNIASATIALVGTAFLSLNIAVNIQSLRSCHSSSESPDLCNYMGSISNGMVSLLLILTLLELCVTISTIAMWCNANCCNSREEISSPPNSV | |
RUO | |
MS4A3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |