missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MSL3L1 Partial ORF (NP_523353.1, 1 a.a. - 79 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00010943-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16133416

  • 4515.00 SEK / 10µg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a nuclear protein and has similarity to drosophila male-specific lethal-3 gene. The drosophila protein plays a critical role in a dosage-compensation pathway, which equalizes X-linked gene expression in males and females. Thus this encoded protein is thought to play a similar function in chromatin remodeling and transcriptional regulation. This gene has been found to undergo X inactivation. There are four alternatively spliced transcript variants of this gene encoding different isoforms. [provided by RefSeq]

Sequence: MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVVVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDEN
Specifications

Specifications

NP_523353.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.43kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVVVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDEN
RUO
MSL3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
10943
MSL3L1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp586J1822/MSL3L1
MSL3
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers