missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MTMR2 Partial ORF (NP_057240, 5 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00008898-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16179605

  • 3343.96 SEK / 10µg

Estimated Shipment: 06-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene is a member of the myotubularin family. The encoded protein possesses phosphatase activity towards phosphatidylinositol-3-phosphate and phosphatidylinositol-3,5-bisphosphate. Mutations in this gene are a cause of Charcot-Marie-Tooth disease type 4B, an autosomal recessive demyelinating neuropathy. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Sequence: SSCESLGSQPAAARPPSVDSLSSASTSHSENSVHTKSASVVSSDSISTSADNFSPDLRVLRESNKLAEMEEPPLLPGENIKDMAKDVTYICPFTGA
Specifications

Specifications

NP_057240
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.3kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
SSCESLGSQPAAARPPSVDSLSSASTSHSENSVHTKSASVVSSDSISTSADNFSPDLRVLRESNKLAEMEEPPLLPGENIKDMAKDVTYICPFTGA
RUO
MTMR2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
8898
MTMR2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CMT4B/CMT4B1/KIAA1073
MTMR2
Yes
Videos
SDS
Documents
Certificates