missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MUTYH Partial ORF (AAH03178, 436 a.a. - 535 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH03178 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4595 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16138031
|
Abnova™
H00004595-Q01.25UG |
25 ug |
6845.00 SEK
25µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16128031
|
Abnova™
H00004595-Q01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a DNA glycosylase involved in oxidative DNA damage repair. The enzyme excises adenine bases from the DNA backbone at sites where adenine is inappropriately paired with guanine, cytosine, or 8-oxo-7,8-dihydroguanine, a major oxidatively damaged DNA lesion. The protein is localized to the nucleus and mitochondria. Mutations in this gene result in heritable predisposition to colon and stomach cancer. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: KLTYQVYGLALEGQTPVTTVPPGARWLTQEEFHTAAVSTAMKKVFRVYQGQQPGTCMGSKRSQVSSPCSRKKPRMGQQVLDNFFRSHISTDAHSLNSAAQSpecifications
AAH03178 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC4416/MYH | |
MUTYH | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
4595 | |
MUTYH (Human) Recombinant Protein (Q01) | |
KLTYQVYGLALEGQTPVTTVPPGARWLTQEEFHTAAVSTAMKKVFRVYQGQQPGTCMGSKRSQVSSPCSRKKPRMGQQVLDNFFRSHISTDAHSLNSAAQ | |
RUO | |
MUTYH | |
Wheat Germ (in vitro) | |
GST | |
Liquid |