missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MYL5 Full-length ORF (AAH40050.1, 1 a.a. - 132 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
4515.00 SEK - 6845.00 SEK
Specifications
Accession Number | AAH40050.1 |
---|---|
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 4636 |
Molecular Weight (g/mol) | 40.26kDa |
Purification Method | Glutathione Sepharose 4 Fast Flow |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16128091
|
Abnova™
H00004636-P01.10UG |
10 ug |
4515.00 SEK
10µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16138091
|
Abnova™
H00004636-P01.25UG |
25μg |
6845.00 SEK
25µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes one of the myosin light chains, a component of the hexameric ATPase cellular motor protein myosin. Myosin is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene product, one of the regulatory light chains, is expressed in fetal muscle and in adult retina, cerebellum, and basal ganglia. [provided by RefSeq]
Sequence: MDQNRDGFIDKEDLKDTYASLGKTNVKDDELDAMLKEASGPINFTMFLNLFGEKLSGTDAEETILNAFKMLDPDGKGKINKEYIKRLLMSQADKMTAEEVDQMFQFASIDVAGNLDYKALSYVITHGEEKEESpecifications
AAH40050.1 | |
4636 | |
Glutathione Sepharose 4 Fast Flow | |
RUO | |
MYL5 | |
Recombinant | |
Liquid |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
40.26kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYL5 | |
Wheat Germ (in vitro) | |
wheat germ expression system |