missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYO9A Full-length ORF (AAH60886.1, 1 a.a. - 729 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004649-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16178101

  • 5030.41 SEK / 25µg

Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: MNINDGGRRRFEDNEHTLRIYPGAISEGTIYCPIPAKKNSTAAEVIESLINKLHLDKTKCYVLAEVKEFGGEEWILNPTDCPVQRMMLWPRMALENRLSGEDYRFLLREKNLDGSIHYGSLQSWLRVTEERRRMMERGFLPQPQQKDFDDLCSLPDLNEKTLLENLRNRFKHEKIYTYVGSILIVINPFKFLPIYNPKYVKMYDNHQLGKLEPHIYAVADVAYHAMLQRKKNQCIVISGESGSGKTQSTNFLIHHLTALSQKGFASGVEQIILGAGPVLEAFGNAKTAHNNNSSRFGKFIQVNYQETGTVLGAYVEKYLLEKSRLVYQEHNERNYHVFYYLLAGASEDERSAFHLKQPEEYHYLNQITKKPLRQSWDDYCYDSEPDCFTVEGEDLRHDFERLQLAMEMVGFLPKTRRQIFSLLSAILHLGNICYKKKTYRDDSIDICNPEVLPIVSELLEVKEEMLFEALVTRKTVTVGEKLILPYKLAEAVTVRNSMAKSLYSALFDWIVFRINHALLNSKDLEHNTKTLSIGVLDIFGFEDYENNSFEQFCINFANERLQHYFNQHIFKLEQEEYRTEGISWHNIDYIDNTCCINLISKKPTGLLHLLDEESNFPQATNQTLLDKFKHQHEDNSYIEFPAVMEPAFIIKHYAGKVKYGVKDFREKNTDHMRPDIVALLRSSKNAFISGMIGIDPVAVFRWAILRAFFRAMVAFREAGKRNIHRKTGK
Specifications

Specifications

AAH60886.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
111.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11061/FLJ13244/MGC71859
MYO9A
Yes
wheat germ expression system
Liquid
4649
MYO9A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNINDGGRRRFEDNEHTLRIYPGAISEGTIYCPIPAKKNSTAAEVIESLINKLHLDKTKCYVLAEVKEFGGEEWILNPTDCPVQRMMLWPRMALENRLSGEDYRFLLREKNLDGSIHYGSLQSWLRVTEERRRMMERGFLPQPQQKDFDDLCSLPDLNEKTLLENLRNRFKHEKIYTYVGSILIVINPFKFLPIYNPKYVKMYDNHQLGKLEPHIYAVADVAYHAMLQRKKNQCIVISGESGSGKTQSTNFLIHHLTALSQKGFASGVEQIILGAGPVLEAFGNAKTAHNNNSSRFGKFIQVNYQETGTVLGAYVEKYLLEKSRLVYQEHNERNYHVFYYLLAGASEDERSAFHLKQPEEYHYLNQITKKPLRQSWDDYCYDSEPDCFTVEGEDLRHDFERLQLAMEMVGFLPKTRRQIFSLLSAILHLGNICYKKKTYRDDSIDICNPEVLPIVSELLEVKEEMLFEALVTRKTVTVGEKLILPYKLAEAVTVRNSMAKSLYSALFDWIVFRINHALLNSKDLEHNTKTLSIGVLDIFGFEDYENNSFEQFCINFANERLQHYFNQHIFKLEQEEYRTEGISWHNIDYIDNTCCINLISKKPTGLLHLLDEESNFPQATNQTLLDKFKHQHEDNSYIEFPAVMEPAFIIKHYAGKVKYGVKDFREKNTDHMRPDIVALLRSSKNAFISGMIGIDPVAVFRWAILRAFFRAMVAFREAGKRNIHRKTGK
RUO
MYO9A
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates