missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human MYST4 Partial ORF (NP_036462, 80 a.a. - 172 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023522-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16177392

  • 3343.96 SEK / 10µg

Estimated Shipment: 14-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: FSSVKPGTFPKSAKGSRGSCNDLRNVDWNKLLRRAIEGLEEPNGSSLKNIEKYLRSQSDLTSTTNNPAFQQRLRLGAKRAVNNGRLLKDGPQY
Specifications

Specifications

NP_036462
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.97kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
FSSVKPGTFPKSAKGSRGSCNDLRNVDWNKLLRRAIEGLEEPNGSSLKNIEKYLRSQSDLTSTTNNPAFQQRLRLGAKRAVNNGRLLKDGPQY
RUO
MYST4
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
23522
MYST4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp313G1618/FLJ90335/KAT6B/KIAA0383/MORF/MOZ2/qkf/querkopf
MYST4
Yes
Videos
SDS
Documents
Certificates