missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ Human NAA20 (aa 92-165) Control Fragment Recombinant Protein
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
Beskrivning
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66574 (PA5-66574. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Catalytic subunit of the NatB complex which catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Asp, Met-Glu, Met-Asn and Met-Gln. Proteins with cell cycle functions are overrepresented in the pool of NatB substrates. Required for maintaining the structure and function of actomyosin fibers and for proper cellular migration.
Specifikationer
Specifikationer
| Tillträdesnummer | P61599 |
| Koncentration | ≥5.0 mg/mL |
| För användning med (applikation) | Blocking Assay, Control |
| Formulering | 1 M urea, PBS with no preservative; pH 7.4 |
| Gen-ID (Entrez) | 51126 |
| Namn | Human NAA20 (aa 92-165) Control Fragment |
| Kvantitet | 100 μL |
| Regulatorisk status | RUO |
| Gene Alias | 1500004D14Rik; 2900026I01Rik; AU041458; D2Ertd186e; dJ1002M8.1; Methionine N-acetyltransferase; N(alpha)-acetyltransferase 20, NatB catalytic subunit; NAA20; N-acetyltransferase 3 homolog; N-acetyltransferase 5; N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae); N-acetyltransferase 5 (GCN5-related, putative); N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, homolog); N-alpha-acetyltransferase 20; N-alpha-acetyltransferase 20, NatB catalytic subunit; NAT3; NAT3P; NAT5; NAT5P; NatB catalytic subunit; natB complex subunit NAT5; N-terminal acetyltransferase B complex catalytic subunit NAA20; N-terminal acetyltransferase B complex catalytic subunit NAT5; N-terminal acetyltransferase complex ARD1 subunit |
| Vanligt namn | NAA20 |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering